Imprimer

Anti-SYN2 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13258-50EA 719 EUR
ABCAAB13258-50
Anti-SYN2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SYN2 (synapsin II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR