Imprimer

Anti-CCNA1 Mouse monoclonal antibody unconjugated [clone: XLA1-3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13337-50EA 691 EUR
ABCAAB13337-50
Anti-CCNA1 Mouse monoclonal antibody unconjugated [clone: XLA1-3]
Anticorps
Type: Primary
Antigen: CCNA1 (cyclin A1)
Clonality: monoclonal
Clone: XLA1-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Xenopus laevis
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR