Imprimer

Anti-Neurokinin A Receptor Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13397-50EA 550 EUR
ABCAAB13397-50
Anti-Neurokinin A Receptor Rabbit polyclonal antibody
Type: Primary
Antigen: Neurokinin A Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR