Imprimer

Anti-PDIA3 Mouse Monoclonal Antibody [clone: MaP.Erp57]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13506-100EA 745 EUR
ABCAAB13506-100
Anti-PDIA3 Mouse Monoclonal Antibody [clone: MaP.Erp57]
Anticorps
Type: Primary
Antigen: PDIA3 (protein disulfide isomerase family A, member 3)
Clonality: monoclonal
Clone: MaP.Erp57
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR