Imprimer

Anti-DLG4 Mouse monoclonal antibody unconjugated [clone: 7E3-1B8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13552-50EA 943 EUR
ABCAAB13552-50
Anti-DLG4 Mouse monoclonal antibody unconjugated [clone: 7E3-1B8]
Anticorps
Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality: monoclonal
Clone: 7E3-1B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR