Imprimer

Anti-CYC Mouse Monoclonal Antibody [clone: 7H8.2C12]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13575-100EA 938 EUR
ABCAAB13575-100
Anti-CYC Mouse Monoclonal Antibody [clone: 7H8.2C12]
Anticorps
Type: Primary
Antigen: CYC (cytochrome C)
Clonality: monoclonal
Clone: 7H8.2C12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR