Imprimer

Anti-ELF5 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13581-100EA 591 EUR
ABCAAB13581-100
Anti-ELF5 Goat polyclonal antibody
Type: Primary
Antigen: ELF5 (E74-like factor 5 (ets domain transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR