Imprimer

Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 59A426]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13597-100EA 735 EUR
ABCAAB13597-100
Anti-TNFAIP3 Mouse Monoclonal Antibody [clone: 59A426]
Anticorps
Type: Primary
Antigen: TNFAIP3 (tumor necrosis factor, alpha-induced protein 3)
Clonality: monoclonal
Clone: 59A426
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR