Imprimer

Anti-Yersinia pestis Mouse monoclonal antibody [clone: 6031]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13639-50EA 550 EUR
ABCAAB13639-50
Anti-Yersinia pestis Mouse monoclonal antibody [clone: 6031]
Type: Primary
Antigen: Yersinia pestis
Clonality: monoclonal
Clone: 6031
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR