Imprimer

Anti-IL25 Mouse monoclonal antibody [clone: 68C1039]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13651-100EA 609 EUR
ABCAAB13651-100
Anti-IL25 Mouse monoclonal antibody [clone: 68C1039]
Type: Primary
Antigen: IL25 (interleukin 25)
Clonality: monoclonal
Clone: 68C1039
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR