Imprimer

Anti-TIRAP Mouse Monoclonal Antibody [clone: 20D1055]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13684-200EA 704 EUR
ABCAAB13684-200
Anti-TIRAP Mouse Monoclonal Antibody [clone: 20D1055]
Anticorps
Type: Primary
Antigen: TIRAP (toll-interleukin 1 receptor (TIR) domain containing adaptor protein)
Clonality: monoclonal
Clone: 20D1055
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR