Imprimer

Anti-TNFRSF12A Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13709-200EA 540 EUR
ABCAAB13709-200
Anti-TNFRSF12A Rabbit polyclonal antibody
Type: Primary
Antigen: TNFRSF12A (tumor necrosis factor receptor superfamily, member 12A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR