Imprimer

Anti-PGLYRP1 Mouse Monoclonal Antibody [clone: 188C424]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13903-100EA 666 EUR
ABCAAB13903-100
Anti-PGLYRP1 Mouse Monoclonal Antibody [clone: 188C424]
Anticorps
Type: Primary
Antigen: PGLYRP1 (peptidoglycan recognition protein 1)
Clonality: monoclonal
Clone: 188C424
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR