Imprimer

Anti-MAPK3 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13909-100EA 540 EUR
ABCAAB13909-100
Anti-MAPK3 Rabbit polyclonal antibody
Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR