Imprimer

Anti-Lamin A + C Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13910-200EA 618 EUR
ABCAAB13910-200
Anti-Lamin A + C Rabbit polyclonal antibody
Type: Primary
Antigen: Lamin A + C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR