Imprimer

Anti-IKBKG Mouse monoclonal antibody [clone: 72C627]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13917-100EA 565 EUR
ABCAAB13917-100
Anti-IKBKG Mouse monoclonal antibody [clone: 72C627]
Type: Primary
Antigen: IKBKG (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma)
Clonality: monoclonal
Clone: 72C627
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR