Imprimer

Anti-D4 GDI Mouse monoclonal antibody [clone: 10D774]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13925-100EA 0 EUR
ABCAAB13925-100
Anti-D4 GDI Mouse monoclonal antibody [clone: 10D774]
Type: Primary
Antigen: D4 GDI
Clonality: monoclonal
Clone: 10D774
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR