Imprimer
Anti-MAP3K7 Rabbit polyclonal antibody
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB13927-100EA 540 EUR
ABCAAB13927-100
Anti-MAP3K7 Rabbit polyclonal antibody
Type: Primary
Antigen: MAP3K7 (mitogen-activated protein kinase kinase kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human