Imprimer

Anti-TLR9 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13928-50EA 540 EUR
ABCAAB13928-50
Anti-TLR9 Rabbit polyclonal antibody
Type: Primary
Antigen: TLR9 (toll-like receptor 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR