Imprimer

Anti-CENPA Mouse monoclonal antibody unconjugated [clone: 3-19]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13939-50EA 746 EUR
ABCAAB13939-50
Anti-CENPA Mouse monoclonal antibody unconjugated [clone: 3-19]
Anticorps
Type: Primary
Antigen: CENPA (centromere protein A)
Clonality: monoclonal
Clone: 42082
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR