Imprimer

Anti-YES1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13954-100EA 550 EUR
ABCAAB13954-100
Anti-YES1 Rabbit polyclonal antibody
Type: Primary
Antigen: YES1 (v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR