Imprimer

Anti-SCGB1A1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB13978-100EA 579 EUR
ABCAAB13978-100
Anti-SCGB1A1 Rabbit polyclonal antibody
Type: Primary
Antigen: SCGB1A1 (secretoglobin, family 1A, member 1 (uteroglobin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR