Imprimer

Anti-NGB Chicken polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14010-50EA 572 EUR
ABCAAB14010-50
Anti-NGB Chicken polyclonal antibody
Type: Primary
Antigen: NGB (neuroglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR