Imprimer

Anti-NFKB1 Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14059-50EA 702 EUR
ABCAAB14059-50
Anti-NFKB1 Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NFKB1 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR