Imprimer

Anti-YWHAQ Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14112-100EA 697 EUR
ABCAAB14112-100
Anti-YWHAQ Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: YWHAQ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR