Imprimer

Anti-14-3-3 sigma Mouse Monoclonal Antibody [clone: 1.N.6]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14123-250EA 739 EUR
ABCAAB14123-250
Anti-14-3-3 sigma Mouse Monoclonal Antibody [clone: 1.N.6]
Anticorps
Type: Primary
Antigen: 14-3-3 sigma (14-3-3 sigma)
Clonality: monoclonal
Clone: 1.N.6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR