Imprimer

Anti-SPON1 Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14271-50EA 787 EUR
ABCAAB14271-50
Anti-SPON1 Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: SPON1 (spondin 1, extracellular matrix protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR