Imprimer

Anti-VAMP2 Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14279-50EA 697 EUR
ABCAAB14279-50
Anti-VAMP2 Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: VAMP2 (vesicle-associated membrane protein 2 (synaptobrevin 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR