Imprimer

Anti-DNMT3A Chicken polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14291-50EA 579 EUR
ABCAAB14291-50
Anti-DNMT3A Chicken polyclonal antibody
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR