Imprimer

Anti-Relaxin 1 + 2 Mouse monoclonal antibody [clone: 8H11kurd]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14331-50EA 565 EUR
ABCAAB14331-50
Anti-Relaxin 1 + 2 Mouse monoclonal antibody [clone: 8H11kurd]
Type: Primary
Antigen: Relaxin 1 + 2
Clonality: monoclonal
Clone: 8H11kurd
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR