Imprimer

Anti-Basal Bodies of Cillia Mouse monoclonal antibody unconjugated [clone: LhS28]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14373-50EA 666 EUR
ABCAAB14373-50
Anti-Basal Bodies of Cillia Mouse monoclonal antibody unconjugated [clone: LhS28]
Anticorps
Type: Primary
Antigen: Basal Bodies of Cillia
Clonality: monoclonal
Clone: LhS28
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR