Imprimer

Anti-BMI1 Mouse Monoclonal Antibody [clone: 1.T.21]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14389-25EA 808 EUR
ABCAAB14389-25
Anti-BMI1 Mouse Monoclonal Antibody [clone: 1.T.21]
Anticorps
Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: monoclonal
Clone: 1.T.21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR