Imprimer

Anti-TP73 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14430-100EA 720 EUR
ABCAAB14430-100
Anti-TP73 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TP73 (tumor protein p73)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR