Imprimer

Anti-AGGF1 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14445-100EA 565 EUR
ABCAAB14445-100
Anti-AGGF1 Rabbit polyclonal antibody
Type: Primary
Antigen: AGGF1 (angiogenic factor with G patch and FHA domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR