Imprimer
Anti-GHRL Rabbit polyclonal antibody
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB14481-500EA 550 EUR
ABCAAB14481-500
Anti-GHRL Rabbit polyclonal antibody
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human