Imprimer

Anti-CCL14 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14534-50EA 565 EUR
ABCAAB14534-50
Anti-CCL14 Rabbit polyclonal antibody
Type: Primary
Antigen: CCL14 (chemokine (C-C motif) ligand 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR