Imprimer

Anti-NDUFA9 Mouse Monoclonal Antibody [clone: 20C11B11B11]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14713-100EA 808 EUR
ABCAAB14713-100
Anti-NDUFA9 Mouse Monoclonal Antibody [clone: 20C11B11B11]
Anticorps
Type: Primary
Antigen: NDUFA9 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa)
Clonality: monoclonal
Clone: 20C11B11B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR