Imprimer

Anti-UQCRFS1 Mouse Monoclonal Antibody [clone: 5A5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14746-100EA 948 EUR
ABCAAB14746-100
Anti-UQCRFS1 Mouse Monoclonal Antibody [clone: 5A5]
Anticorps
Type: Primary
Antigen: UQCRFS1 (ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1)
Clonality: monoclonal
Clone: 5A5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR