Imprimer

Anti-FOXM1 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14758-100EA 361 EUR
ABCAAB14758-100
Anti-FOXM1 Goat polyclonal antibody
Type: Primary
Antigen: FOXM1 (forkhead box M1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR