Imprimer

Anti-S100 Mouse Monoclonal Antibody [clone: 4B3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14849-250EA 739 EUR
ABCAAB14849-250
Anti-S100 Mouse Monoclonal Antibody [clone: 4B3]
Anticorps
Type: Primary
Antigen: S100
Clonality: monoclonal
Clone: 4B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR