Imprimer

Anti-LYVE1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB14917-100EA 775 EUR
ABCAAB14917-100
Anti-LYVE1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: LYVE1 (lymphatic vessel endothelial hyaluronan receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR