Imprimer

Anti-BCAP31 Rat monoclonal antibody [clone: CC-4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15045-50EA 593 EUR
ABCAAB15045-50
Anti-BCAP31 Rat monoclonal antibody [clone: CC-4]
Type: Primary
Antigen: BCAP31 (B-cell receptor-associated protein 31)
Clonality: monoclonal
Clone: CC-4
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR