Imprimer

Anti-Erythromycin Mouse monoclonal antibody [clone: 1.BB.893]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15061-200EA 565 EUR
ABCAAB15061-200
Anti-Erythromycin Mouse monoclonal antibody [clone: 1.BB.893]
Type: Primary
Antigen: Erythromycin
Clonality: monoclonal
Clone: 1.BB.893
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR