Imprimer

Anti-Novobiocin Mouse monoclonal antibody [clone: 3H349]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15069-100EA 540 EUR
ABCAAB15069-100
Anti-Novobiocin Mouse monoclonal antibody [clone: 3H349]
Type: Primary
Antigen: Novobiocin
Clonality: monoclonal
Clone: 3H349
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR