Imprimer

Anti-Streptomycin Mouse monoclonal antibody unconjugated [clone: 1.BB.944]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15071-200EA 0 EUR
ABCAAB15071-200
Anti-Streptomycin Mouse monoclonal antibody unconjugated [clone: 1.BB.944]
Type: Primary
Antigen: Streptomycin
Clonality: monoclonal
Clone: 1.BB.944
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR