Imprimer

Anti-COX2 / Cyclooxygenase 2 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15191-500EA 749 EUR
ABCAAB15191-500
Anti-COX2 / Cyclooxygenase 2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: COX2 / Cyclooxygenase 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR