Imprimer

Anti-Melanoma gp100 Mouse monoclonal antibody [clone: SPM142]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15229-7EA 550 EUR
ABCAAB15229-7
Anti-Melanoma gp100 Mouse monoclonal antibody [clone: SPM142]
Type: Primary
Antigen: Melanoma gp100
Clonality: monoclonal
Clone: SPM142
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR