Imprimer

Anti-Myogenin Mouse monoclonal antibody [clone: SPM144]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15232-500EA 586 EUR
ABCAAB15232-500
Anti-Myogenin Mouse monoclonal antibody [clone: SPM144]
Type: Primary
Antigen: Myogenin
Clonality: monoclonal
Clone: SPM144
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR