Imprimer

Anti-Ceftiofur Mouse monoclonal antibody [clone: 1.BB.880]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15255-250EA 565 EUR
ABCAAB15255-250
Anti-Ceftiofur Mouse monoclonal antibody [clone: 1.BB.880]
Type: Primary
Antigen: Ceftiofur
Clonality: monoclonal
Clone: 1.BB.880
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR