Imprimer

Anti-Growth Hormone Mouse Monoclonal Antibody [clone: SPM106]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15318-7EA 579 EUR
ABCAAB15318-7
Anti-Growth Hormone Mouse Monoclonal Antibody [clone: SPM106]
Type: Primary
Antigen: Growth Hormone
Clonality: Monoclonal
Clone: SPM106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR